Anti-CD4 Rabbit Polyclonal Antibody

Supplier: Proteintech
19068-1-AP-150UL
10084-348EA 402 USD
10084-348
Anti-CD4 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Thymus Tissue, 1:500-1:5000

Type: Primary
Antigen: CD4
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR