Anti-Brevican Rabbit Polyclonal Antibody

Supplier: Proteintech
19017-1-AP-150UL
10083-680EA 402 USD
10083-680
Anti-Brevican Rabbit Polyclonal Antibody
Antibodies
Brevican is a brain-specific proteoglycan belonging to the aggrecan family.

Western Blot: Human Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: Brevican
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR