Anti-NOX2 Rabbit Polyclonal Antibody

Supplier: Proteintech
19013-1-AP-150UL
10091-228EA 537.7 USD
10091-228
Anti-NOX2 Rabbit Polyclonal Antibody
Antibodies
NOX2, also named as CYBB, CGD, 91-phox, gp91-1, gp91-phox, p22 phagocyte B-cytochrome, cytochrome b-245 and beta polypeptide, is a critical component of the membrane-bound oxidase of phagocytes that generates superoxide. It is the terminal component of a respiratory chain that transfers single electrons from cytoplasmic NADPH across the plasma membrane to molecular oxygen on the exterior. This full length protein has three glycosylation sites. CYBB is found in human cardiomyocytes as multiple bands:the signal between 55 and 65 kDa is probably the unglycosylated protein, because the predicted molecular weight of unglycosylated CYBB protein is approximately 58 kDa to 65 kDa in phagocytes and the bands around 80 kDa probably represent glycosylated CYBB, as has also been shown in human umbilical vein endothelial cells. In other reports, it also can be detected a band of 91 kDa. In our results, it reavels the bands of 55-65 kDa and 80 kDa besides a band of 140 kDa that we do not find the reason.

Western Blot: Mouse Brain Tissue, 1:200-1:2000

Type: Primary
Antigen: NOX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR