Anti-PIF1 Rabbit Polyclonal Antibody

Supplier: Proteintech
19006-1-AP-150UL
10092-242EA 402 USD
10092-242
Anti-PIF1 Rabbit Polyclonal Antibody
Antibodies
PIF1, also named as ATP-dependent DNA helicase PIF1, is a 641 amino acid protein, which belongs to the helicase family. PIF1 as a DNA-dependent ATPase and 5'-3' DNA helicase is required for the maintenance of both mitochondrial and nuclear genome stability. PIF1 localizes in the nucleus and is involved in the maintenance of telomeric DNA. PIF1 exists as four isoforms. isoform 1 is a 70 kDa protein and isoform 2 is 63 kDa protein.

Western Blot: Mouse Thymus Tissue, 1:200-1:2000

Type: Primary
Antigen: PIF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR