Anti-ZFP36L2 Rabbit Polyclonal Antibody

Supplier: Proteintech
19005-1-AP-150UL
10097-268EA 402 USD
10097-268
Anti-ZFP36L2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HepG2 Cells, 1:200-1:2000

Type: Primary
Antigen: ZFP36L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR