Anti-PTH Rabbit Polyclonal Antibody

Supplier: Proteintech
19001-1-AP-150UL
10093-036EA 402 USD
10093-036
Anti-PTH Rabbit Polyclonal Antibody
Antibodies
PTH belongs to the parathyroid hormone family. PTH elevates calcium level by dissolving the salts in bone and preventing their renal excretion. Defects in PTH are a cause of familial isolated hypoparathyroidism (FIH).

Western Blot: Recombinant Protein, 1:1000-1:10000

Type: Primary
Antigen: PTH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR