Anti-POU3F1 Rabbit Polyclonal Antibody

Supplier: Proteintech
18997-1-AP-150UL
10091-542EA 402 USD
10091-542
Anti-POU3F1 Rabbit Polyclonal Antibody
Antibodies
OCT6, also named as POU3F1 and OTF6, belongs to the POU transcription factor family and Class-3 subfamily. OCT6 is a transcription factor that binds to the octamer motif (5'-ATTTGCAT-3'). It is thought to be involved in early embryogenesis and neurogenesis. Tha antibody is specific to OCT6.

Western Blot: HEK-293 Cells, 1:200-1:2000

Type: Primary
Antigen: POU3F1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR