Anti-OCT2 Rabbit Polyclonal Antibody

Supplier: Proteintech
18996-1-AP-150UL
10091-536EA 524.48 USD
10091-536
Anti-OCT2 Rabbit Polyclonal Antibody
Antibodies
OCT2, also named as POU2F2 and OTF2, belongs to the POU transcription factor family and Class-2 subfamily. OCT2 is a transcription factor that specifically binds to the octamer motif (5'-ATTTGCAT-3'). OCT2 regulates transcription in a number of tissues in addition to activating immunoglobulin gene expression. It modulates transcription transactivation by NR3C1, AR and PGR. Isoform 5 activates the U2 small nuclear RNA (snRNA) promoter. The antibody is special to OCT2.

Western Blot: Mouse Thymus Tissue, 1:500-1:5000; IHC: Human spleen Tissue, 1:20-1:200

Type: Primary
Antigen: OCT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR