Anti-SLC24A4 Rabbit Polyclonal Antibody

Supplier: Proteintech
18992-1-AP-150UL
10094-730EA 402 USD
10094-730
Anti-SLC24A4 Rabbit Polyclonal Antibody
Antibodies
SLC24A4, also named as NCKX4, belongs to the sodium/potassium/calcium exchanger family. SLC24A4 transports 1 Ca2+ and 1 K+ in exchange for 4 Na+. Genetic variations in SLC24A4 are associated with skin/hair/eye pigmentation type 6 (SHEP6).

Western Blot: Mouse Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: SLC24A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR