Anti-HAUS3 Rabbit Polyclonal Antibody

Supplier: Proteintech
18991-1-AP-150UL
10088-054EA 402 USD
10088-054
Anti-HAUS3 Rabbit Polyclonal Antibody
Antibodies
HAUS3, also named as C4orf15, belongs to the HAUS3 family. It is Required for mitotic centrosome and spindle integrity. HAUS3 is a component of the HAUS augmin-like complex which regulates mitotic spindle assembly and centrosome integrity and is required for completion of cytokinesis. The complex interacts with the gamma-tubulin ring complex and this interaction is required for spindle assembly.

Western Blot: HepG2 Cells, 1:500-1:5000; IHC: Human Liver cancer Tissue, 1:20-1:200

Type: Primary
Antigen: HAUS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR