Anti-EGFR Rabbit Polyclonal Antibody

Supplier: Proteintech
18986-1-AP-150UL
10086-312EA 453 USD
10086-312
Anti-EGFR Rabbit Polyclonal Antibody
Antibodies
ERBB1, also mamed as EGFR, is a receptor for EGF, but also for other members of the EGF family, as TGF-alpha, amphiregulin, betacellulin, heparin-binding EGF-like growth factor, GP30 and vaccinia virus growth factor. ERBB1 is involved in the control of cell growth and differentiation. Phosphorylates MUC1 in breast cancer cells and increases the interaction of MUC1 with C-SRC and CTNNB1/beta-catenin. Isoform 2/truncated isoform of ERBB1 may act as an antagonist. EGFR is associated with lung cancer. The antibody is specific to isoform1.

Western Blot: HeLa Cells, 1:500-1:5000; IF: Hela Cells, 1:10-1:100; IP: MCF-7 Cells, 1:200-1:2000

Type: Primary
Antigen: EGFR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR