Anti-iNOS Rabbit Polyclonal Antibody

Supplier: Proteintech
18985-1-AP-150UL
10088-914EA 537.7 USD
10088-914
Anti-iNOS Rabbit Polyclonal Antibody
Antibodies
NOS2, also named as iNOS and NOS2A, produces nitric oxide (NO) which is a messenger molecule with diverse functions throughout the body. In macrophages, NO mediates tumoricidal and bactericidal actions. NO is a reactive free radical which acts as a biologic mediator in several processes, including neurotransmission and antimicrobial and antitumoral activities. NOS2 is a nitric oxide synthase which is expressed in liver and is inducible by a combination of lipopolysaccharide and certain cytokines. This antibody is specific to NOS2.

IHC: Human Liver, 1:20-1:200

Type: Primary
Antigen: iNOS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR