Anti-TXK Rabbit Polyclonal Antibody

Supplier: Proteintech
18981-1-AP-150UL
10096-522EA 402 USD
10096-522
Anti-TXK Rabbit Polyclonal Antibody
Antibodies
TXK, also named as PTK4, is an enzyme that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation.

IHC: Human Lymphoma, 1:20-1:200; IF: Hela, 1:10-1:100

Type: Primary
Antigen: TXK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR