Anti-SH3PXD2A Rabbit Polyclonal Antibody

Supplier: Proteintech
18976-1-AP-150UL
10094-544EA 402 USD
10094-544
Anti-SH3PXD2A Rabbit Polyclonal Antibody
Antibodies
SH3PXD2A is also named as FISH, KIAA0418, SH3MD1, TKS5. SH3PXD2A contains an amino-terminal PX domain followed by five SH3 domains. It is a cytoplasmic protein in normal fibroblasts.The p140 and p130 forms of SH3PXD2A may be generated by other splice variations. The complexity of SH3PXD2A isoforms is also evident in different cell types. For example in human platelets a single band of 150kDa was detected, whereas all three forms were found in human fibroblasts and vascular smooth muscle cells.This antibody is specific to SH3PXD2A.

Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human breast cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: SH3PXD2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR