Anti-RTP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
18973-1-AP-150UL
10094-044EA 453 USD
10094-044
Anti-RTP1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HEK-293 Cells, 1:200-1:2000

Type: Primary
Antigen: RTP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR