Anti-NECAB3 Rabbit Polyclonal Antibody

Supplier: Proteintech
18940-1-AP-150UL
10090-998EA 402 USD
10090-998
Anti-NECAB3 Rabbit Polyclonal Antibody
Antibodies
NECAB3, also named as APBA2BP, NIP1, SYTIP2 and XB51, is a potential substrate of Nek2. NECAB3 inhibits the interaction of APBA2 with beta-amyloid precursor protein (APP). NECAB3 has three isoforms with MW 40 kDa, 44 kDa and 22 kDa.

Western Blot: Human Brain Tissue, 1:200-1:2000; IHC: Human pancreas Tissue, 1:20-1:200

Type: Primary
Antigen: NECAB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR