Anti-SOX13 Rabbit Polyclonal Antibody

Supplier: Proteintech
18902-1-AP-150UL
10095-016EA 524.48 USD
10095-016
Anti-SOX13 Rabbit Polyclonal Antibody
Antibodies
SRY-BOX 13(SOX13), identified a sequence homologous more than 60% to the SRY HMG box ,with a centrally located HMG box and an N-terminal leucine zipper motif with a neighboring glutamine stretch called a Q box. SOX13 binds to the consensus HMG-box motif. It's an IDDM-specific human autoantigen, and a gamma-delta-specific gene in the immune system. It can modulates Wnt/TCF activity. This is a rabbit polyclonal raised against part of N-terminal chain of SOX13 of human origin.

Western Blot:Jurkat Cells, 1:1000-1:10000; IHC: Human Kidney, 1:20-1:200; IF: Hela, 1:20-1:200

Type: Primary
Antigen: SOX13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR