Anti-CEP170L Rabbit Polyclonal Antibody

Supplier: Proteintech
18899-1-AP-150UL
10084-644EA 402 USD
10084-644
Anti-CEP170L Rabbit Polyclonal Antibody
Antibodies
IF: Hela Cells, 1:10-1:100; Western Blot: Mouse Testis Tissue, 1:500-1:5000

Type: Primary
Antigen: CEP170L
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR