Anti-NEURL Rabbit Polyclonal Antibody

Supplier: Proteintech
18898-1-AP-150UL
10091-050EA 537.7 USD
10091-050
Anti-NEURL Rabbit Polyclonal Antibody
Antibodies
NEURL(Neuralized-like protein 1A) is also named as NEURL1, NEURL1A, RNF67.Neurl1 is a RING-dependent E3 ubiquitin ligase that can regulate Notch signaling in mammals and it expression promotes lysosomal degradation of Jagged1 in cells and blocks signaling from Jagged1-expressing cells to neighboring Notch-expressing cells. Neurl-dependent turnover of Jagged1 and inhibition of Notch signaling depends upon N-terminal myristoylation and plasma membrane localization of Neurl1.This antibody is specific to NEURL.

Western Blot: Mouse Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: NEURL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR