Anti-ADH1C Rabbit Polyclonal Antibody

Supplier: Proteintech
18897-1-AP-150UL
10082-522EA 402 USD
10082-522
Anti-ADH1C Rabbit Polyclonal Antibody
Antibodies
ADH1C(Alcohol dehydrogenase 1C) is also named as ADH3 and belongs to the zinc-containing alcohol dehydrogenase family.It catalyzes the rate-limiting step for ethanol metabolism: the oxidation of alcohol to acetaldehyde.The ADH1C gene is active in intestine and kidney in fetal and early postnatal life, and persists in the stomach and liver in adult life.

Western Blot: Mouse Liver Tissue, 1:1000-1:10000; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: ADH1C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR