Anti-LAPTM4B Rabbit Polyclonal Antibody

Supplier: Proteintech
18895-1-AP-150UL
10089-510EA 402 USD
10089-510
Anti-LAPTM4B Rabbit Polyclonal Antibody
Antibodies
LAPTM4B, a novel oncoprotein, is associated with the prognosis of cancer, and may play important roles in the initiation, promotion and metastasis of tumors. It was initially identified as a hepatocellular carcinoma-associated gene, and was recently shown to be up-regulated in various human cancers. Some studies indicated that LAPTM4B may be an useful marker of prognosis in ovarian carcinoma and endometrial cancer. LAPTM4B belongs to the LAPTM4/LAPTM5 transporter family. It has 3 isoforms with molecular masses 41kd, 35 kd and 24 kd. This antibody can recognize all the 3 isoforms of LAPTM4B.

Western Blot: Mouse lymph Tissue, 1:500-1:5000; IHC: Human Liver cancer Tissue, 1:20-1:200

Type: Primary
Antigen: LAPTM4B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR