Anti-PTRF Rabbit Polyclonal Antibody

Supplier: Proteintech
18892-1-AP-150UL
10092-338EA 402 USD
10092-338
Anti-PTRF Rabbit Polyclonal Antibody
Antibodies
Polymerase I and transcript release factor(PTRF) is an essential factor in the biogenesis of caveolae, which involve in numerous process, such as signal transduction and membrane and lipid trafficking. PTRF can terminate transciption of RNA Pol I, which involves pausing of transcription by TTF1, and dissocaition of the transcription complex, release the complex from the template.

Western Blot:A549 Cells, 1:1000-1:10000; IHC: Human pancreas Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100; IP: Mouse lung Tissue, 1:200-1:2000

Type: Primary
Antigen: PTRF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR