Anti-UTP3 Rabbit Polyclonal Antibody

Supplier: Proteintech
18886-1-AP-150UL
10096-744EA 402 USD
10096-744
Anti-UTP3 Rabbit Polyclonal Antibody
Antibodies
Western Blot:A549 Cells, 1:1000-1:10000; IF: HepG2, 1:20-1:200

Type: Primary
Antigen: UTP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR