Anti-UBR2 Rabbit Polyclonal Antibody

Supplier: Proteintech
18853-1-AP-150UL
10096-690EA 402 USD
10096-690
Anti-UBR2 Rabbit Polyclonal Antibody
Antibodies
UBR2(Ubiquitin-protein ligase E3-alpha-2) is also named as C6orf133, KIAA0349 and N-recognin-2. It is a component of the N-end rule pathway and recognizes and binds to proteins bearing specific N-terminal residues that are destabilizing according to the N-end rule, leading to their ubiquitination and subsequent degradation. It has four isoforms produced by alternative splicing.

Western Blot:BxPC-3 Cells, 1:200-1:2000; IF: HepG2 Cells, 1:10-1:100; IHC: Human pancreas Tissue, 1:20-1:200

Type: Primary
Antigen: UBR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR