Anti-ENTPD8 Rabbit Polyclonal Antibody

Supplier: Proteintech
18848-1-AP-150UL
10081-294EA 402 USD
10081-294
Anti-ENTPD8 Rabbit Polyclonal Antibody
Antibodies
ENTPD8, also named as UNQ2492 and PRO5779, belongs to the GDA1/CD39 NTPase family. ENTPD8 is a canalicular ectonucleoside NTPDase which responsible for the main hepatic NTPDase activity. Ectonucleoside NTPDases catalyze the hydrolyzis of gamma- and beta-phosphate residues of nucleotides, playing a central role in concentration of extracellular nucleotides. Has activity toward ATP, ADP, UTP and UDP, but not toward AMP. ENTPD8 catalyze the reaction :ATP + 2 H2O = AMP + 2 phosphate.

Western Blot: HepG2 Cells, 1:500-1:5000

Type: Primary
Antigen: ENTPD8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR