Anti-IRF2BP2 Rabbit Polyclonal Antibody

Supplier: Proteintech
18847-1-AP-150UL
10088-966EA 402 USD
10088-966
Anti-IRF2BP2 Rabbit Polyclonal Antibody
Antibodies
IRF2BP2 belongs to the IRF2BP family. IRF2BP2 acts as a transcriptional repressor and as a transcriptional corepressor in a IRF2-dependent manner. This repression is not mediated at least in part by histone deacetylase activities.

Western Blot:Jurkat Cells, 1:1000-1:10000; IHC: Human lung cancer Tissue, 1:20-1:200; IP:K-562 Cells, 1:200-1:2000

Type: Primary
Antigen: IRF2BP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR