Anti-VN1R4 Rabbit Polyclonal Antibody

Supplier: Proteintech
18846-1-AP-150UL
10096-992EA 402 USD
10096-992
Anti-VN1R4 Rabbit Polyclonal Antibody
Antibodies
VN1R4, also named as V1RL4, belongs to the G-protein coupled receptor 1 family. VN1R4 is a pheromone receptor.

Western Blot: HeLa Cells, 1:200-1:2000

Type: Primary
Antigen: VN1R4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR