Anti-RAB12 Rabbit Polyclonal Antibody

Supplier: Proteintech
18843-1-AP-150UL
10093-130EA 402 USD
10093-130
Anti-RAB12 Rabbit Polyclonal Antibody
Antibodies
RAB12 belongs to the small GTPase superfamily and Rab family. RAB12 locates in Golgi apparatus membrane.

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human Liver, 1:20-1:200

Type: Primary
Antigen: RAB12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR