Anti-USP32 Rabbit Polyclonal Antibody

Supplier: Proteintech
18838-1-AP-150UL
10096-864EA 402 USD
10096-864
Anti-USP32 Rabbit Polyclonal Antibody
Antibodies
USP32 belongs to the peptidase C19 family. USP32 catalytic the reaction :Ubiquitin C-terminal thioester + H2O = ubiquitin + a thiol.

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human pancreas, 1:20-1:200

Type: Primary
Antigen: USP32
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR