Anti-CD36 Rabbit Polyclonal Antibody

Supplier: Proteintech
18836-1-AP-150UL
10084-340EA 524.48 USD
10084-340
Anti-CD36 Rabbit Polyclonal Antibody
Antibodies
CD36, also named as GP3B and GP4, seems to have numerous potential physiological functions. CD36 binds to collagen, thrombospondin, anionic phospholipids, long chain fatty acids and may function in the transport and/or as a regulator of fatty acid transport.and oxidized LDL. CD36 may function as a cell adhesion molecule. It mediates cytoadherence of Plasmodium falciparum parasitized erythrocytes. Mutation of CD36 will cause platelet glycoprotein IV deficiency which known as CD36 deficiency. Genetic variations in CD36 are associated with susceptibility to coronary heart disease type 7 (CHDS7). This antibody 18836-1-AP is raised against a synthesized peptide of 100-116 amino acids of CD36.

Western Blot: RAW 264.7 Cells, 1:200-1:2000; IHC: Human spleen Tissue, 1:20-1:200; FC: RAW 264.7 Cells, N/A

Type: Primary
Antigen: CD36
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR