Anti-DRIM Rabbit Polyclonal Antibody

Supplier: Proteintech
18830-1-AP-150UL
10086-132EA 402 USD
10086-132
Anti-DRIM Rabbit Polyclonal Antibody
Antibodies
UTP20, also named as DRIM, interacts with KIAA0649.

Western Blot: Mouse ovary Tissue, 1:500-1:5000; IF: Hela Cells, 1:20-1:200

Type: Primary
Antigen: DRIM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR