Anti-USP34 Rabbit Polyclonal Antibody

Supplier: Proteintech
18827-1-AP-150UL
10096-866EA 402 USD
10096-866
Anti-USP34 Rabbit Polyclonal Antibody
Antibodies
USP34, also named as KIAA0570 and KIAA0729, belongs to the peptidase C19 family. USP34 recognizes and hydrolyzes the peptide bond at the C-terminal Gly of ubiquitin. It is involved in the processing of poly-ubiquitin precursors as well as that of ubiquinated proteins. The antibody recognize all these 3 isoform of USP34.

Western Blot: Human Brain Tissue, 1:500-1:5000; IHC: Human breast cancer Tissue, 1:20-1:200

Type: Primary
Antigen: USP34
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR