Anti-RBP2 Rabbit Polyclonal Antibody

Supplier: Proteintech
18825-1-AP-150UL
10093-440EA 402 USD
10093-440
Anti-RBP2 Rabbit Polyclonal Antibody
Antibodies
KDM5A, also named as JARID1A, RBBP2 and RBP2, belongs to the JARID1 histone demethylase family. KDM5A is a Histone demethylase that specifically demethylates 'Lys-4' of histone H3, thereby playing a central role in histone code. Does not demethylate histone H3 'Lys-9', H3 'Lys-27', H3 'Lys-36', H3 'Lys-79' or H4 'Lys-20'. RBP2 may stimulate transcription mediated by nuclear receptors. It may be involved in transcriptional regulation of Hox proteins during cell differentiation. It may participate in transcriptional repression of cytokines such as CXCL12

Western Blot: HeLa Cells, 1:200-1:2000

Type: Primary
Antigen: RBP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR