Anti-MLN Rabbit Polyclonal Antibody

Supplier: Proteintech
18824-1-AP-150UL
10090-278EA 402 USD
10090-278
Anti-MLN Rabbit Polyclonal Antibody
Antibodies
MLN, also named as MAP, belongs to the motilin family. MLN plays an important role in the regulation of interdigestive gastrointestinal motility and indirectly causes rhythmic contraction of duodenal and colonic smooth muscle.

Western Blot: Human Stomach Tissue, 1:1000-1:10000

Type: Primary
Antigen: MLN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR