Anti-CCNI2 Rabbit Polyclonal Antibody

Supplier: Proteintech
18822-1-AP-150UL
10084-220EA 402 USD
10084-220
Anti-CCNI2 Rabbit Polyclonal Antibody
Antibodies
CCNI2 belongs to the cyclin family.

Western Blot: Human Brain Tissue, 1:500-1:5000; IHC: Human lung cancer, 1:20-1:200

Type: Primary
Antigen: CCNI2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR