Anti-LILRA5 Rabbit Polyclonal Antibody

Supplier: Proteintech
18821-1-AP-150UL
10089-414EA 402 USD
10089-414
Anti-LILRA5 Rabbit Polyclonal Antibody
Antibodies
LILRA5, also named as ILT11, LILRB7, LIR9 and CD85f, may plays a role in triggering innate immune responses. It does not function in class I MHC antigens recognition. The antibody recognizes the isoform 1 and isoform 2 of LILRA5.

Western Blot:K-562 Cells, 1:500-1:5000

Type: Primary
Antigen: LILRA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR