Anti-GPR142 Rabbit Polyclonal Antibody

Supplier: Proteintech
18820-1-AP-150UL
10087-778EA 524.48 USD
10087-778
Anti-GPR142 Rabbit Polyclonal Antibody
Antibodies
GPR142, also named as PGR2, belongs to the G-protein coupled receptor 1 family. It is an orphan receptor. The antibody recognize the 51kd GRP142.

Western Blot:SH-SY5Y Cells, 1:200-1:2000

Type: Primary
Antigen: GPR142
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR