Anti-OTOP3 Rabbit Polyclonal Antibody

Supplier: Proteintech
18819-1-AP-150UL
10091-634EA 402 USD
10091-634
Anti-OTOP3 Rabbit Polyclonal Antibody
Antibodies
OTOP3 belongs to the otopetrin family.

Western Blot: MCF7 Cells, 1:200-1:2000

Type: Primary
Antigen: OTOP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR