Anti-RAB41 Rabbit Polyclonal Antibody

Supplier: Proteintech
18818-1-AP-150UL
10093-208EA 402 USD
10093-208
Anti-RAB41 Rabbit Polyclonal Antibody
Antibodies
RAB41 belongs to the small GTPase superfamily and Rab family.

Western Blot:PC-3 Cells, 1:1000-1:10000

Type: Primary
Antigen: RAB41
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR