Anti-PSKH2 Rabbit Polyclonal Antibody

Supplier: Proteintech
18813-1-AP-150UL
10092-884EA 402 USD
10092-884
Anti-PSKH2 Rabbit Polyclonal Antibody
Antibodies
PSKH2 belongs to the protein kinase superfamily and CAMK Ser/Thr protein kinase family. PSKH2 catalytic the reaction: ATP + a protein = ADP + a phosphoprotein.

Western Blot: HepG2 Cells, 1:1000-1:10000; IHC: Human Kidney, 1:20-1:200

Type: Primary
Antigen: PSKH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR