Anti-INO80 Rabbit Polyclonal Antibody

Supplier: Proteintech
18810-1-AP-150UL
10088-912EA 537.7 USD
10088-912
Anti-INO80 Rabbit Polyclonal Antibody
Antibodies
INO80, also named as INOC1 and KIAA1259, belongs to the SNF2/RAD54 helicase family. The INO80 complex remodels chromatin by shifting nucleosomes. INO80 recruited by YY1 to YY1-activated INO80. In vitro, INO80 has double stranded DNA-dependent ATPase activity

Western Blot: Mouse Liver Tissue, 1:200-1:2000

Type: Primary
Antigen: INO80
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR