Anti-GCHFR Rabbit Polyclonal Antibody

Supplier: Proteintech
18809-1-AP-150UL
10087-434EA 402 USD
10087-434
Anti-GCHFR Rabbit Polyclonal Antibody
Antibodies
GCHFR,also named as p35 and GFRP, belongs to the GFRP family. GCHFR mediates tetrahydrobiopterin inhibition of GTP cyclohydrolase 1. This inhibition is reversed by L-phenylalanine.

Western Blot: Mouse skin Tissue, 1:500-1:5000; IHC: Human cervix, 1:20-1:200

Type: Primary
Antigen: GCHFR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR