Anti-TSPAN11 Rabbit Polyclonal Antibody

Supplier: Proteintech
18807-1-AP-150UL
10096-422EA 402 USD
10096-422
Anti-TSPAN11 Rabbit Polyclonal Antibody
Antibodies
TSPAN11 belongs to the tetraspanin (TM4SF) family.

Western Blot: Human Brain Tissue, 1:200-1:2000

Type: Primary
Antigen: TSPAN11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR