Anti-MEPE Rabbit Polyclonal Antibody

Supplier: Proteintech
18804-1-AP-150UL
10081-626EA 402 USD
10081-626
Anti-MEPE Rabbit Polyclonal Antibody
Antibodies
Matrix extracellular phosphoglycoprotein (MEPE), also known as osteoblast/osteocyte factor 45 (OF45), is amember of the small integrin-binding ligand N-linked glycopro-tein (SIBLING) family . MEPE promotes renal phosphate excretion and modulates mineralization . It can be phosphorylated by secreted protein kinases. . MEPE is expressed in osteoblasts, osteocytes and odontoblasts, and the expression has also been reported in numerous human tumor cell lines . It is a 525-amino acid protein with a calculated molecular weight of 58 kDa. This antibody raised against a synthetic peptide (CTHGRKYHYVPHRQNNSTR) detects a major band (43 kDa) and two minor bands (98 kDa and 34 kDa).

Western Blot: HepG2 Cells, 1:1000-1:10000

Type: Primary
Antigen: MEPE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR