Anti-PCDHA3 Rabbit Polyclonal Antibody

Supplier: Proteintech
18803-1-AP-150UL
10091-922EA 402 USD
10091-922
Anti-PCDHA3 Rabbit Polyclonal Antibody
Antibodies
PCDHA3 is a potential calcium-dependent cell-adhesion protein. It may be involved in the establishment and maintenance of specific neuronal connections in the brain. The antibody can recognize both the isoforms of PCDHA3.

Western Blot:A549 Cells, 1:200-1:2000; IHC: Human Brain Tissue, 1:20-1:200

Type: Primary
Antigen: PCDHA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR