Anti-INTS8 Rabbit Polyclonal Antibody

Supplier: Proteintech
18802-1-AP-150UL
10088-936EA 402 USD
10088-936
Anti-INTS8 Rabbit Polyclonal Antibody
Antibodies
INTS8, also named as C8orf52, is a Component of the Integrator complex which is a complex involved in the small nuclear RNAs (snRNA) U1 and U2 transcription and in their 3'-box-dependent processing. INTS8 is associated with the C-terminal domain (CTD) of RNA polymerase II largest subunit (POLR2A) and is recruited to the U1 and U2 snRNAs genes.

Western Blot: MCF7 Cells, 1:200-1:1000; IF: Hela Cells, 1:10-1:100

Type: Primary
Antigen: INTS8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR