Anti-HOXD13 Rabbit Polyclonal Antibody

Supplier: Proteintech
18736-1-AP-150UL
10088-418EA 402 USD
10088-418
Anti-HOXD13 Rabbit Polyclonal Antibody
Antibodies
HOXD13, also named as HOX4I, belongs to the Abd-B homeobox family. HOXD13 is a sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis. Mutation of HOXD13 will cause synpolydactyly (SPD), brachydactyly type D (BDD), syndactyly type 5brachydactyly-syndactyly syndrome (BDSD) or brachydactyly type E (BDE).Present polyclonal anti-HOX13D antibody(18736-1-AP) is produced by immunizing animals with part of N-terminus chain of HOX13D and detect a 36-kDa band in cell and tissues.

Western Blot:A549 Cells, 1:1000-1:10000; IHC: Human Brain, 1:20-1:200

Type: Primary
Antigen: HOXD13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR