Anti-HOXD12 Rabbit Polyclonal Antibody

Supplier: Proteintech
18735-1-AP-150UL
10088-416EA 402 USD
10088-416
Anti-HOXD12 Rabbit Polyclonal Antibody
Antibodies
HOXD12, also named as HOX4H, belongs to the Abd-B homeobox family. HOXD12 is a sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis. The exact role of this gene has not been determined.

Western Blot: Mouse Liver Tissue, 1:200-1:2000

Type: Primary
Antigen: HOXD12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR