Anti-HOXB1 Rabbit Polyclonal Antibody

Supplier: Proteintech
18732-1-AP-150UL
10088-412EA 402 USD
10088-412
Anti-HOXB1 Rabbit Polyclonal Antibody
Antibodies
HOXB1, also named as HOX2I, belongs to the Antp homeobox family and the Labial subfamily. HOXB1 is sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis. HOXB1 acts on the anterior body structures. Establishment of the spatial colinearity in the embryo is directly controlled by the Hox genes themselves. 18732-1-AP is a rabbit polyclonal antibody raised against a peptide mapping within an internal region of human.

Western Blot: Mouse Liver Tissue, 1:200-1:1000

Type: Primary
Antigen: HOXB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR