Anti-GPX5 Rabbit Polyclonal Antibody

Supplier: Proteintech
18731-1-AP-150UL
10087-822EA 402 USD
10087-822
Anti-GPX5 Rabbit Polyclonal Antibody
Antibodies
GPX5 belongs to the glutathione peroxidase family. GPX5 protects cells and enzymes from oxidative damage, by catalyzing the reduction of hydrogen peroxide, lipid peroxides and organic hydroperoxide, by glutathione. Catalytic the reaction: 2 glutathione + H2O2 = glutathione disulfide + 2 H2O. GPX5 constitute a glutathionine peroxidase-like protective system against peroxide damage in sperm membrane lipids.

Western Blot: Human Testis Tissue, 1:200-1:2000; IHC: Human Testis, 1:20-1:200

Type: Primary
Antigen: GPX5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR